Learn More
Eurofins Discoverx PathHunter Human OPRD1 (Opioid Delta) Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455518
Encompass
The PathHunter Human OPRD1 (Opioid Delta) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure OPRD1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55518 Supplier Eurofins Discoverx Supplier No. 7930673C3
The PathHunter Human OPRD1 (Opioid Delta) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure OPRD1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.