Learn More
Eurofins Discoverx PathHunter Human SSTR2 Activated GPCR Internalization Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456069
Encompass
The PathHunter Human SSTR2 Stable Cell Line Activated GPCR Internalization Assay (CHO-K1) contains a stable clonal cell line used to measure SSTR2 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56069 Supplier Eurofins Discoverx Supplier No. 7930676C2
The PathHunter Human SSTR2 Stable Cell Line Activated GPCR Internalization Assay (CHO-K1) contains a stable clonal cell line used to measure SSTR2 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.