Learn More
Eurofins Discoverx PathHunter Human SDF-1 alpha (CXCR4) β-Arrestin Cell-Based Assay Kit (C2C12)
Catalog No. 5000455582
Encompass
The PathHunter Human SDF-1 alpha (CXCR4) β-Arrestin Stable Cell Line Assay (C2C12) contains a stable clonal cell line used to measure CXCR4 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55582 Supplier Eurofins Discoverx Supplier No. 7930203C7
The PathHunter Human SDF-1 alpha (CXCR4) β-Arrestin Stable Cell Line Assay (C2C12) contains a stable clonal cell line used to measure CXCR4 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.