Learn More
Eurofins Discoverx PathHunter Human PRLR-JAK2 Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455742
Encompass
The PathHunter Human PRLR-JAK2 Stable Cell Line Functional Kinase Assay (U2OS) measures PRLR cytokine receptor activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55742 Supplier Eurofins Discoverx Supplier No. 7930687C3
The PathHunter Human PRLR-JAK2 Stable Cell Line Functional Kinase Assay (U2OS) measures PRLR cytokine receptor activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.