Learn More
Eurofins Discoverx PathHunter Mouse MIP-3 alpha (CCR6) β-Arrestin Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455895
Encompass
The PathHunter Mouse MIP-3 alpha (CCR6) β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure mCCR6 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55895 Supplier Eurofins Discoverx Supplier No. 7930694C2
The PathHunter Mouse MIP-3 alpha (CCR6) β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure mCCR6 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.