Learn More
Eurofins Discoverx PathHunter Human GLP-2 (GLP-2 Receptor) Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455470
Encompass
The PathHunter Human GLP-2 (GLP2R) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure GLP2R (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55470 Supplier Eurofins Discoverx Supplier No. 7930750C3
The PathHunter Human GLP-2 (GLP2R) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure GLP2R (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.