Learn More
Eurofins Discoverx PathHunter Human GHR-JAK1 Functional Kinase Cell-Based Assay Kit (U2OS)
Catalog No. 5000455963
Encompass
The PathHunter Human GHR-JAK1 Stable Cell Line Functional Kinase Assay (U2OS) measures GHR cytokine receptor activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55963 Supplier Eurofins Discoverx Supplier No. 7930756C3
The PathHunter Human GHR-JAK1 Stable Cell Line Functional Kinase Assay (U2OS) measures GHR cytokine receptor activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.