Learn More
Eurofins Discoverx PathHunter Human p53 (TP53) Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000455524
Encompass
The PathHunter Human p53 (TP53) Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure p53 DNA damage pathway activation via nuclear translocation of p53. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55524 Supplier Eurofins Discoverx Supplier No. 7930757C3
The PathHunter Human p53 (TP53) Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure p53 DNA damage pathway activation via nuclear translocation of p53. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.