Learn More
Eurofins Discoverx PathHunter Human MR Nuclear Receptor Protein Interaction Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456075
Encompass
The PathHunter Human MR Nuclear Receptor Stable Cell Line Protein Interaction Assay (CHO-K1) contains a stable clonal cell line used to measure MR nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56075 Supplier Eurofins Discoverx Supplier No. 7930758C2
The PathHunter Human MR Nuclear Receptor Stable Cell Line Protein Interaction Assay (CHO-K1) contains a stable clonal cell line used to measure MR nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.