Learn More
Eurofins Discoverx PathHunter Human Complement C5a (C5AR1) Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455472
Encompass
The PathHunter Human Complement C5a (C5AR1) Stable Cell Line Activated Internalization Assay (U2OS) contains a stable clonal cell line used to measure C5AR1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55472 Supplier Eurofins Discoverx Supplier No. 7930777C3
The PathHunter Human Complement C5a (C5AR1) Stable Cell Line Activated Internalization Assay (U2OS) contains a stable clonal cell line used to measure C5AR1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.