Learn More
Eurofins Discoverx PathHunter Human CDC25A Degradation Cell-Based Assay Kit (NCI-H460)
Catalog No. 5000456077
Encompass
The PathHunter Human CDC25A Degradation Stable Cell Line Assay (NCI-H460) measures pathway activation-induced degradation of CDC25A target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56077 Supplier Eurofins Discoverx Supplier No. 7930776C12
The PathHunter Human CDC25A Degradation Stable Cell Line Assay (NCI-H460) measures pathway activation-induced degradation of CDC25A target protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.