Learn More
Eurofins Discoverx PathHunter Rat DRD2L β-Arrestin Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455526
Encompass
The PathHunter Rat DRD2L β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure DRD2L (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55526 Supplier Eurofins Discoverx Supplier No. 7930785C2
The PathHunter Rat DRD2L β-Arrestin Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure DRD2L (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.