Learn More
Eurofins Discoverx PathHunter Human S1P1 (EDG1) Total GPCR Internalization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455786
Encompass
The PathHunter Human S1P1 (EDG1) Stable Cell Line Total GPCR Internalization Assay (HEK 293) contains a stable clonal cell line used to measure EDG1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55786 Supplier Eurofins Discoverx Supplier No. 7930784C1
The PathHunter Human S1P1 (EDG1) Stable Cell Line Total GPCR Internalization Assay (HEK 293) contains a stable clonal cell line used to measure EDG1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.