Learn More
Eurofins Discoverx PathHunter Human NPR1 Functional Kinase Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455487
Encompass
The HitHunter Human NPR1 cGMP Stable Cell Line Assay (CHO-K1) measures NPR1 pathway activation via an increase in cGMP levels. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55487 Supplier Eurofins Discoverx Supplier No. 7930804C2
The HitHunter Human NPR1 cGMP Stable Cell Line Assay (CHO-K1) measures NPR1 pathway activation via an increase in cGMP levels. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.