Learn More
Eurofins Discoverx PathHunter Human Insulin Receptor B (INSRb) Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455966
Encompass
The PathHunter Human Insulin Receptor B (INSRb) Stable Cell Line Internalization Assay (U2OS) measures activation-induced internalization of the receptor tyrosine kinase, INSRb via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55966 Supplier Eurofins Discoverx Supplier No. 7930808C3
The PathHunter Human Insulin Receptor B (INSRb) Stable Cell Line Internalization Assay (U2OS) measures activation-induced internalization of the receptor tyrosine kinase, INSRb via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.