Learn More
Eurofins Discoverx PathHunter Human Glucagon (GCG Receptor) Total GPCR Internalization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455527
Encompass
The PathHunter Human Glucagon (GCGR) Stable Cell Line Total GPCR Internalization Assay (HEK 293) contains a stable clonal cell line used to measure GCGR (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55527 Supplier Eurofins Discoverx Supplier No. 7930820C1
Add to Cart
Add to Cart
The PathHunter Human Glucagon (GCGR) Stable Cell Line Total GPCR Internalization Assay (HEK 293) contains a stable clonal cell line used to measure GCGR (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.