Learn More
Eurofins Discoverx PathHunter Human Keap1-NRF2 Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000456093
Encompass
The PathHunter Human Keap1-NRF2 Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure Keap1-NRF2 pathway activation via nuclear translocation of NRF2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56093 Supplier Eurofins Discoverx Supplier No. 7930821C3
The PathHunter Human Keap1-NRF2 Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure Keap1-NRF2 pathway activation via nuclear translocation of NRF2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.