Learn More
Eurofins Discoverx PathHunter Human SREBP2 Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000455787
Encompass
The PathHunter Human SREBP2 Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure cholesterol biosynthesis pathway activation via nuclear translocation of SREBP2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55787 Supplier Eurofins Discoverx Supplier No. 7930830C3
The PathHunter Human SREBP2 Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure cholesterol biosynthesis pathway activation via nuclear translocation of SREBP2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.