Learn More
Eurofins Discoverx PathHunter Human TORC1 (CRTC1) Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000455191
Encompass
The PathHunter Human TORC1 (CRTC1) Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure CRTC/CREB pathway activation via nuclear translocation of TORC1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55191 Supplier Eurofins Discoverx Supplier No. 7930833C3
The PathHunter Human TORC1 (CRTC1) Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure CRTC/CREB pathway activation via nuclear translocation of TORC1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.