Learn More
Eurofins Discoverx PathHunter Human MDC (CCR4) Total GPCR Internalization Cell-Based Assay Kit (U2OS)
Supplier:  Eurofins Discoverx 7930845C3
Encompass
The PathHunter Human MDC (CCR4) Stable Cell Line Total GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CCR4 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55260