Learn More
Eurofins Discoverx PathHunter Human MTNR1A Total GPCR Internalization Cell-Based Assay Kit (DLD1)
Catalog No. 5000455332
Encompass
The PathHunter Human MTNR1A Stable Cell Line Total GPCR Internalization Assay (DLD1) contains a stable clonal cell line used to measure MTNR1A (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55332 Supplier Eurofins Discoverx Supplier No. 7930847C13
The PathHunter Human MTNR1A Stable Cell Line Total GPCR Internalization Assay (DLD1) contains a stable clonal cell line used to measure MTNR1A (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.