Learn More
Eurofins Discoverx PathHunter Human PR beta Nuclear Receptor Protein Interaction Cell-Based Assay Kit (U2OS)
Catalog No. 5000455259
Encompass
The PathHunter Human PR beta Nuclear Receptor Stable Cell Line Protein Interaction Assay (U2OS) contains a stable clonal cell line used to measure activated PRβ nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55259 Supplier Eurofins Discoverx Supplier No. 7930851C3
Add to Cart
Add to Cart
The PathHunter Human PR beta Nuclear Receptor Stable Cell Line Protein Interaction Assay (U2OS) contains a stable clonal cell line used to measure activated PRβ nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.