Learn More
Eurofins Discoverx PathHunter Human CHRM1 β-Arrestin Cell-Based Assay Kit (U2OS)
Catalog No. 5000455158
Encompass
The PathHunter Human CHRM1 β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure CHRM1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55158 Supplier Eurofins Discoverx Supplier No. 7930859C3
The PathHunter Human CHRM1 β-Arrestin Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure CHRM1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.