Learn More
Eurofins Discoverx PathHunter Human PR alpha Nuclear Receptor Protein Interaction Cell-Based Assay Kit (U2OS)
Catalog No. 5000455276
Encompass
The PathHunter Human PR alpha Nuclear Receptor Stable Cell Line Protein Interaction Assay (U2OS) contains a stable clonal cell line used to measure activated PRα nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55276 Supplier Eurofins Discoverx Supplier No. 7930852C3
The PathHunter Human PR alpha Nuclear Receptor Stable Cell Line Protein Interaction Assay (U2OS) contains a stable clonal cell line used to measure activated PRα nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.