Learn More
Eurofins Discoverx PathHunter Human LXR beta-NCoR1 Nuclear Receptor Protein Interaction Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455331
Encompass
The PathHunter Human LXR beta-NCoR1 Nuclear Receptor Stable Cell Line Protein Interaction Assay (HEK 293) contains a stable clonal cell line used to measure activated LXRβ-NCoR1 nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55331 Supplier Eurofins Discoverx Supplier No. 7930856C1
The PathHunter Human LXR beta-NCoR1 Nuclear Receptor Stable Cell Line Protein Interaction Assay (HEK 293) contains a stable clonal cell line used to measure activated LXRβ-NCoR1 nuclear hormone receptor interaction with SRCPs. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.