Learn More
Eurofins Discoverx PathHunter Human XBP1 Nuclear Translocation Cell-Based Assay Kit (U2OS)
Supplier:  Eurofins Discoverx 7930857C3
Encompass
The PathHunter Human XBP1 Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure the Endoplasmic Reticulum stress response via nuclear translocation of XBP1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55372