Learn More
Eurofins Discoverx PathHunter Human FOXO3 Nuclear Translocation Cell-Based Assay Kit (U2OS)
Catalog No. 5000455196
Encompass
The PathHunter Human FOXO3 Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure PI3K pathway activation via nuclear translocation of FOXO3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55196 Supplier Eurofins Discoverx Supplier No. 7930876C3
The PathHunter Human FOXO3 Stable Cell Line Nuclear Translocation Assay (U2OS) contains a stable clonal cell line used to measure PI3K pathway activation via nuclear translocation of FOXO3. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.