Learn More
Eurofins Discoverx PathHunter Human OPRM1 (Opioid mu) β-Arrestin-1 Cell-Based Assay Kit (U2OS)
Catalog No. 5000455197
Encompass
The PathHunter Human OPRM1 (Opioid mu) β-Arrestin-1 Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure OPRM1 (GPCR) activity via recruitment of β-Arrestin-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55197 Supplier Eurofins Discoverx Supplier No. 7930878C3
The PathHunter Human OPRM1 (Opioid mu) β-Arrestin-1 Stable Cell Line Assay (U2OS) contains a stable clonal cell line used to measure OPRM1 (GPCR) activity via recruitment of β-Arrestin-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.