Learn More
Eurofins Discoverx PathHunter Human FGR Kinase Activity Cell-Based Assay Kit (U2OS)
Catalog No. 5000455305
Encompass
The PathHunter Human FGR Stable Cell Line Kinase Activity Assay (U2OS) measures FGR CTK activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55305 Supplier Eurofins Discoverx Supplier No. 7930884C3
The PathHunter Human FGR Stable Cell Line Kinase Activity Assay (U2OS) measures FGR CTK activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.