Learn More
Eurofins Discoverx PathHunter Human ADCYAP1R1 (PAC1) Total GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455262
Encompass
The PathHunter Human ADCYAP1R1 (PAC1) Stable Cell Line Total GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure ADCYAP1R1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55262 Supplier Eurofins Discoverx Supplier No. 7930885C3
The PathHunter Human ADCYAP1R1 (PAC1) Stable Cell Line Total GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure ADCYAP1R1 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.