Learn More
Eurofins Discoverx PathHunter Human TGFBR1/TGFBR2 Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455199
Encompass
The PathHunter Human TGFBR1/TGFBR2 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of TGFBR1/TGFBR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55199 Supplier Eurofins Discoverx Supplier No. 7930889C3
The PathHunter Human TGFBR1/TGFBR2 Dimerization Stable Cell Line Assay (U2OS) measures ligand-induced dimerization of TGFBR1/TGFBR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.