Learn More
Eurofins Discoverx PathHunter Human CXCR4 Total GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455160
Encompass
The PathHunter Human CXCR4 Stable Cell Line Total GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CXCR4 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55160 Supplier Eurofins Discoverx Supplier No. 7930890C3
The PathHunter Human CXCR4 Stable Cell Line Total GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CXCR4 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.