Learn More
Eurofins Discoverx PathHunter Human FLT3 Kinase Activity Cell-Based Assay Kit (U2OS)
Catalog No. 5000455306
Encompass
The PathHunter Human FLT3 Stable Cell Line Kinase Activity Assay (U2OS) measures FLT3 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55306 Supplier Eurofins Discoverx Supplier No. 7930901C3
The PathHunter Human FLT3 Stable Cell Line Kinase Activity Assay (U2OS) measures FLT3 receptor tyrosine kinase activity via ligand-induced SH2 domain protein recruitment. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.