Learn More
Eurofins Discoverx PathHunter Human BMP-7 (ACVR2/ACVR1) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455307
Encompass
The PathHunter Human BMP-7 (ACVR2/ACVR1) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ACVR1/ACVR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55307 Supplier Eurofins Discoverx Supplier No. 7930906C3
The PathHunter Human BMP-7 (ACVR2/ACVR1) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ACVR1/ACVR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.