Learn More
Eurofins Discoverx PathHunter Human PTGER4 β-Arrestin Cell-Based Assay Kit (BHK-21)
Catalog No. 5000455200
Encompass
The PathHunter Human PTGER4 β-Arrestin Stable Cell Line Assay (BHK-21) contains a stable clonal cell line used to measure rPTGER4 (GPCR) activity via recruitment of β-Arrestin2. Rat protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55200 Supplier Eurofins Discoverx Supplier No. 7930908C16
The PathHunter Human PTGER4 β-Arrestin Stable Cell Line Assay (BHK-21) contains a stable clonal cell line used to measure rPTGER4 (GPCR) activity via recruitment of β-Arrestin2. Rat protein. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.