Learn More
Eurofins Discoverx PathHunter Human LXR alpha Nuclear Translocation Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000456047
Encompass
The PathHunter Human LXR alpha Stable Cell Line Nuclear Translocation Assay (CHO-K1) contains a stable clonal cell line used to measure LXRα nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-56047 Supplier Eurofins Discoverx Supplier No. 7930215C2
The PathHunter Human LXR alpha Stable Cell Line Nuclear Translocation Assay (CHO-K1) contains a stable clonal cell line used to measure LXRα nuclear hormone receptor activation via nuclear translocation. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.