Learn More
Eurofins Discoverx PathHunter Human IL-17A (IL17RA/IL17RC) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455341
Encompass
The PathHunter Human IL-17A (IL17RA/IL17RC) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL17RA/IL17RC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55341 Supplier Eurofins Discoverx Supplier No. 7930931C1
The PathHunter Human IL-17A (IL17RA/IL17RC) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL17RA/IL17RC. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.