Learn More
Eurofins Discoverx PathHunter Human IL-8 (CXCR2) Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455429
Encompass
The PathHunter Human IL-8 (CXCR2) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CXCR2 (IL8RB) (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55429 Supplier Eurofins Discoverx Supplier No. 7930934C3
The PathHunter Human IL-8 (CXCR2) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CXCR2 (IL8RB) (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.