Learn More
Eurofins Discoverx PathHunter Human Heregulin beta 2 (ErbB4/ErbB4) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455232
Encompass
The PathHunter Human Heregulin beta 2 (ErbB4/ErbB4) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ErbB4/ErbB4. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55232 Supplier Eurofins Discoverx Supplier No. 7930961C3
The PathHunter Human Heregulin beta 2 (ErbB4/ErbB4) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ErbB4/ErbB4. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.