Learn More
Eurofins Discoverx PathHunter Human Heregulin beta 1 (ErbB2/ErbB4) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455344
Encompass
The PathHunter Human Heregulin beta 1 (ErbB2/ErbB4) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ErbB2/ErbB4. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55344 Supplier Eurofins Discoverx Supplier No. 7930960C3
The PathHunter Human Heregulin beta 1 (ErbB2/ErbB4) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ErbB2/ErbB4. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.