Learn More
Eurofins Discoverx PathHunter Human BMP-9 (ACVRL1/ACVR2) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455205
Encompass
The PathHunter Human BMP-9 (ACVRL1/ACVR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ACVRL1/ACVR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55205 Supplier Eurofins Discoverx Supplier No. 7930963C3
The PathHunter Human BMP-9 (ACVRL1/ACVR2) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of ACVRL1/ACVR2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.