Learn More
Eurofins Discoverx PathHunter Human Erythropoietin (EpoR/EpoR) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455267
Encompass
The PathHunter Human Erythropoietin (EpoR/EpoR) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of EpoR/EpoR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55267 Supplier Eurofins Discoverx Supplier No. 7930965C3
The PathHunter Human Erythropoietin (EpoR/EpoR) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of EpoR/EpoR. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.