Learn More
Eurofins Discoverx PathHunter Human MIP-3α (CCR6) Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455343
Encompass
The PathHunter Human MIP-3α (CCR6) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CCR6 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55343 Supplier Eurofins Discoverx Supplier No. 7930966C3
The PathHunter Human MIP-3α (CCR6) Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure CCR6 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.