Learn More
Eurofins Discoverx PathHunter Human GPR35 Activated GPCR Internalization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455216
Encompass
The PathHunter Human GPR35 Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure GPR35 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55216 Supplier Eurofins Discoverx Supplier No. 7930968C3
Add to Cart
Add to Cart
The PathHunter Human GPR35 Stable Cell Line Activated GPCR Internalization Assay (U2OS) contains a stable clonal cell line used to measure GPR35 (GPCR) internalization via early-endosome localization. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.