Learn More
Eurofins Discoverx PathHunter Human IL-3 (IL3RA/CD131) Dimerization Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455258
Encompass
The PathHunter Human IL-3 (IL3RA/CD131) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL3RA/CD131. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55258 Supplier Eurofins Discoverx Supplier No. 7930969C1
The PathHunter Human IL-3 (IL3RA/CD131) Stable Cell Line Dimerization Assay (HEK 293) measures ligand-induced dimerization of IL3RA/CD131. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.