Learn More
Eurofins Discoverx PathHunter Mouse Glucagon (GCG Receptor) β-Arrestin-1 Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455215
Encompass
The PathHunter Mouse Glucagon (GCGR) β-Arrestin-1 Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GCGR (GPCR) activity via recruitment of β-Arrestin-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55215 Supplier Eurofins Discoverx Supplier No. 7930973C2
The PathHunter Mouse Glucagon (GCGR) β-Arrestin-1 Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GCGR (GPCR) activity via recruitment of β-Arrestin-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.