Learn More
Eurofins Discoverx PathHunter Human IL-5 (IL5RA/CSF2RB) Dimerization Cell-Based Assay Kit (U2OS)
Catalog No. 5000455303
Encompass
The PathHunter Human IL-5 (IL5RA/CSF2RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL5RA/CSF2RB. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55303 Supplier Eurofins Discoverx Supplier No. 7930972C3
The PathHunter Human IL-5 (IL5RA/CSF2RB) Stable Cell Line Dimerization Assay (U2OS) measures ligand-induced dimerization of IL5RA/CSF2RB. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.