Learn More
Eurofins Discoverx PathHunter Human FFAR1 (GPR40) β-Arrestin-1 Cell-Based Assay Kit (HEK 293)
Catalog No. 5000455155
Encompass
The PathHunter Human FFAR1 (GPR40) β-Arrestin-1 Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure FFAR1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55155 Supplier Eurofins Discoverx Supplier No. 7930977C1
The PathHunter Human FFAR1 (GPR40) β-Arrestin-1 Stable Cell Line Assay (HEK 293) contains a stable clonal cell line used to measure FFAR1 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.