Learn More
Eurofins Discoverx PathHunter Human GPR120S β-Arrestin-1 Cell-Based Assay Kit (CHO-K1)
Catalog No. 5000455217
Encompass
The PathHunter Human GPR120S β-Arrestin-1 Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GPR120S (GPCR) activity via recruitment of β-Arrestin-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55217 Supplier Eurofins Discoverx Supplier No. 7930978C2
The PathHunter Human GPR120S β-Arrestin-1 Stable Cell Line Assay (CHO-K1) contains a stable clonal cell line used to measure GPR120S (GPCR) activity via recruitment of β-Arrestin-1. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.