Learn More
Eurofins Discoverx PathHunter Mouse PTGER4 β-Arrestin Cell-Based Assay Kit (DLD1)
Catalog No. 5000455285
Encompass
The PathHunter Mouse PTGER4 β-Arrestin Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure mPTGER4 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.
Catalog No. 50-004-55285 Supplier Eurofins Discoverx Supplier No. 7930982C13
The PathHunter Mouse PTGER4 β-Arrestin Stable Cell Line Assay (DLD1) contains a stable clonal cell line used to measure mPTGER4 (GPCR) activity via recruitment of β-Arrestin2. This is a complete kit that includes the cell vials and all required reagents needed for cell culture and running the assay.